Learn More
Methyl 7-bromoheptanoate, 98%
Product Code. 11342239
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342239 25 g 25g
11332239 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11342239 Supplier Thermo Scientific Alfa Aesar Supplier No. H27234.14
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
CAS: 54049-24-0 | C8H15BrO2 | 223.11 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54049-24-0
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD02258672
InChI Key BXRLUWIDTDLHQE-UHFFFAOYSA-N
Synonym methyl-7-bromoheptanoate, 7-bromoheptanoic acid methyl ester, heptanoic acid, 7-bromo-, methyl ester, acmc-1aya7, 13-ethyl-17-ethynyl-17-hydroxy-11-methylidene-2,6,7,8,9,10,12,13,14,15,16,17-dodecahydro-1h-cyclopenta a phenanthren-3-one
PubChem CID 554086
IUPAC Name methyl 7-bromoheptanoate
SMILES COC(=O)CCCCCCBr
Specifications
Density 1.257
Quantity 25 g
Formula Weight 223.11
Percent Purity 98%
Chemical Name or Material Methyl 7-bromoheptanoate