Learn More
Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate, TRC
Product Code. 30465547
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30465547 Supplier Toronto Research Chemicals Supplier No. TRCM30509350MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate
CAS 62140-67-4
Molecular Formula C11H14O5S
Quantity 50 mg
InChI Formula InChI=1S/C11H14O5S/c1-4-17(13,14)8-5-6-10(15-2)9(7-8)11(12)16-3/h5-7H,4H2,1-3H3
SMILES CCS(=O)(=O)c1ccc(OC)c(c1)C(=O)OC
IUPAC Name methyl 5-ethylsulfonyl-2-methoxybenzoate
Molecular Weight (g/mol) 258.29
Formula Weight 258.0562