Learn More
Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 5-(Ethylsulfonyl)-2-methoxybenzoate |
| CAS | 62140-67-4 |
| Molecular Formula | C11H14O5S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C11H14O5S/c1-4-17(13,14)8-5-6-10(15-2)9(7-8)11(12)16-3/h5-7H,4H2,1-3H3 |
| SMILES | CCS(=O)(=O)c1ccc(OC)c(c1)C(=O)OC |
| IUPAC Name | methyl 5-ethylsulfonyl-2-methoxybenzoate |
| Molecular Weight (g/mol) | 258.29 |
| Formula Weight | 258.0562 |