Learn More
Methyl 5-Bromovalerate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 5-Bromovalerate |
| CAS | 5454-83-1 |
| Molecular Formula | C6H11BrO2 |
| Quantity | 10 g |
| Synonym | 5-Bromopentanoic Acid Methyl Ester, 5-Bromovaleric Acid Methyl Ester, Methyl 5-Bromopentanoate, Methyl delta-Bromovalerate, Methyl ω-Bromopentanoate, NSC 23228 |
| InChI Formula | InChI=1S/C6H11BrO2/c1-9-6(8)4-2-3-5-7/h2-5H2,1H3 |
| SMILES | COC(=O)CCCCBr |
| IUPAC Name | methyl 5-bromopentanoate |
| Molecular Weight (g/mol) | 195.05 |
| Formula Weight | 193.9942 |