Learn More
Methyl (5-((2-methyl-4-oxopentan-2-yl)thio)-1H-benzo[d]imidazol-2-yl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl N-[5-[(2-Methyl-4-oxopentan-2-yl)sulfanyl]-1H-benzimidazol-2-yl]carbamate |
| CAS | 2469260-71-5 |
| Analyte or Component Names | Methyl N-[5-[(2-Methyl-4-oxopentan-2-yl)sulfanyl]-1H-benzimidazol-2-yl]carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N3 O3 S |
| Synonym | Methyl N-[5-[(2-Methyl-4-oxopentan-2-yl)sulfanyl]-1H-benzimidazol-2-yl]carbamate, Methyl [5-[(2-Methyl-4-oxopentan-2-yl)sulfanyl]-1H-benzimidazol-2-yl]carbamate, Albendazole related, GSK 2269758A |
| InChI Formula | InChI=1S/C15H19N3O3S/c1-9(19)8-15(2,3)22-10-5-6-11-12(7-10)17-13(16-11)18-14(20)21-4/h5-7H,8H2,1-4H3,(H2,16,17,18,20) |
| Solution Type | Neat |
| SMILES | COC(=O)Nc1nc2cc(SC(C)(C)CC(=O)C)ccc2[nH]1 |
| IUPAC Name | methyl N-[5-(2-methyl-4-oxopentan-2-yl)sulfanyl-1H-benzimidazol-2-yl]carbamate |
| Show More |