Learn More
Methyl 4-hydroxy-2-hexynoate, 97%, Thermo Scientific Chemicals
Catalog Number 11450568
Click to view available options
Quantity:
1 g
5 g
25 g
112780-04-8
C7H10O3
142.154
MFCD04039770
GMESNUCOCRRYJV-UHFFFAOYSA-N
methyl 4-hydroxy-2-hexynoate, 4-hydroxy-2-hexynoic acid methyl ester, acmc-20akge, methyl-4-hydroxy-2-hexynoate, 4-hydroxy-hex-2-ynoic acid methyl ester, 2-hexynoic acid,4-hydroxy-, methyl ester
13859766
methyl 4-hydroxyhex-2-ynoate
CCC(C#CC(=O)OC)O
112780-04-8
C7H10O3
142.154
MFCD04039770
GMESNUCOCRRYJV-UHFFFAOYSA-N
methyl 4-hydroxy-2-hexynoate, 4-hydroxy-2-hexynoic acid methyl ester, acmc-20akge, methyl-4-hydroxy-2-hexynoate, 4-hydroxy-hex-2-ynoic acid methyl ester, 2-hexynoic acid,4-hydroxy-, methyl ester
13859766
methyl 4-hydroxyhex-2-ynoate
CCC(C#CC(=O)OC)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Linear Formula CH3CH2CH(OH)C≡CCO2CH3
Refractive Index 1.462
Quantity 1 g
Formula Weight 142.15
Percent Purity 97%
Chemical Name or Material Methyl 4-hydroxy-2-hexynoate