Learn More
Methyl 4-Formylbenzoate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 1571-08-0 |
| Melting Point | 56°C to 59°C |
| Density | 1.2 g/cm3 at 20°C |
| Boiling Point | 264°C to 266°C/1013 hPa |
| Molecular Formula | 4-(OHC)C6H4COOCH3 |
| MDL Number | MFCD00006950 |
| Quantity | 100 g |
| Synonym | Methyl 4-formylbenzoate, 4-Formylbenzoic acid methyl ester, Terephthalaldehydic acid methyl ester, Methyl 4-formylbenzoate |
| Molecular Weight (g/mol) | 164.16 |
| Grade | Research |