missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobenzoate, 98+%

Product Code. 11422927
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422927 25 g 25g
11412927 5 g 5g
11432927 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422927 Supplier Thermo Scientific Alfa Aesar Supplier No. A15517.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-42-1 | C8H7BrO2 | 215.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl 4-bromobenzoate is used as a starting material in the synthesis of three-carbon-bridged 5-substituted furo[2,3-d]pyrimidine and 6-substituted pyrrolo[2,3-d]pyrimidine and methyl 4-tri-n-butylstannylbenzoate. It is also used to stimulate microbial dechlorination of polychlorinated biphenyls. Further, it is used in the preparation of stable radioiodinating reagent to label monoclonal antibodies for radiotherapy of cancer.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-42-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00013531
InChI Key CZNGTXVOZOWWKM-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
PubChem CID 12081
IUPAC Name methyl 4-bromobenzoate
SMILES COC(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 77°C to 81°C
Density 1.69
Quantity 25 g
Beilstein 2045132
Merck Index 14,1408
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material Methyl 4-bromobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.