missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(thien-2-yl)acrylate, 97%, Thermo Scientific™

Product Code. p-7024913
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459606 1 g 1g
11469606 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459606 Supplier Thermo Scientific Alfa Aesar Supplier No. B20177.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20883-96-9
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00173772
InChI Key HKVOGMDMMCLQFJ-SNAWJCMRSA-N
Synonym methyl 3-2-thienyl acrylate, methyl 3-thien-2-yl acrylate, methyl 3-thien-2-yl-acrylate, methyl e-3-thiophen-2-ylprop-2-enoate, methyl 2e-3-thiophen-2-yl prop-2-enoate, methyl 3-thiophen-2-yl prop-2-enoate, thiophene-2-acrylic acid methyl ester, e-methyl 3-thiophen-2-yl acrylate, methyl 2e-3-2-thienyl-2-propenoate, methyl 2e-3-2-thienyl prop-2-enoate
PubChem CID 5709013
IUPAC Name methyl (E)-3-thiophen-2-ylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CS1

Specifications

Melting Point 48°C to 49°C
Quantity 1 g
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material Methyl 3-(thien-2-yl)acrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.