Learn More
Methyl 3-Phenylpropionate(3-Phenylpropionic Acid Methyl Ester), TRC
Product Code. 30422615
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30422615 100 g
30428753 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30422615 Supplier Toronto Research Chemicals Supplier No. TRCP336190100G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-25-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
Synonym Benzenepropanoic Acid Methyl Ester, 3-Phenylpropanoic Acid Methyl Ester, 3-Phenylpropionic Acid Methyl Ester, Dihydrocinnamic Acid Methyl Ester, Methyl 3-phenylpropanoate, Methyl benzenepropanoate, Methyl dihydrocinnamate, Methyl Hydrocinnamate, Methyl beta-Phenylpropionate, NSC 10128, NSC 404188, beta-Phenylpropionic Acid Methyl Ester
IUPAC Name methyl 3-phenylpropanoate
SMILES COC(=O)CCc1ccccc1
Specifications
Quantity 100 g
InChI Formula InChI=1S/C10H12O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-6H,7-8H2,1H3
Formula Weight 164.0837
Chemical Name or Material Methyl 3-Phenylpropionate(3-Phenylpropionic Acid Methyl Ester)