Learn More
Methyl 3-formyl-4-hydroxybenzoate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 24589-99-9 |
| Molecular Formula | C9H8O4 |
| MDL Number | MFCD01661473 |
| Quantity | 1 g |
| SMILES | COC(=O)C1=CC(C=O)=C(O)C=C1 |
| Molecular Weight (g/mol) | 180.16 |
| Percent Purity | 97% |
| Chemical Name or Material | Methyl 3-formyl-4-hydroxybenzoate |