missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl 3-chlorosulfonylthiophene-2-carboxylate, 95%

Product Code. p-7040683
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15465816

Brand: Alfa Aesar™ H60521.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 59337-92-7
Molecular Formula C6H5ClO4S2
Molecular Weight (g/mol) 240.67
MDL Number MFCD00068160
InChI Key PJVJBDAUWILEOG-UHFFFAOYSA-N
Synonym methyl 3-chlorosulfonyl thiophene-2-carboxylate, 2-carbomethoxy-3-thiophenesulfonyl chloride, methylchlorosulfonylthiophenecarboxylate, methyl 3-chlorosulfonyl-2-thiophenecarboxylate, 3-chlorosulfonyl-2-thiophene carboxylic acid methyl ester, methyl-3-chlorosulfonylthiophene-2-carboxylate, 2-methoxycarbonyl thiophene-3-sulfonyl chloride, 2-thiophenecarboxylic acid, 3-chlorosulfonyl-, methyl ester, 2-methoxycarbonyl-3-thiophenesulfonyl chloride, 2-methoxycarbonyl thiophene-3-sulphonyl chloride
PubChem CID 2736733
IUPAC Name methyl 3-(chlorosulfonyl)thiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)S(Cl)(=O)=O

Specifications

Melting Point 61°C to 64°C
Quantity 1 g
UN Number UN3261
Formula Weight 240.68
Percent Purity 95%
Chemical Name or Material Methyl 3-chlorosulfonylthiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.