missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Catalog Number p-7046379
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3395-91-3
C4H7BrO2
167.00
MFCD00000250
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
76934
methyl 3-bromopropanoate
COC(=O)CCBr
This item is not returnable. View return policy
3395-91-3
C4H7BrO2
167.00
MFCD00000250
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
methyl 3-bromopropionate, propanoic acid, 3-bromo-, methyl ester, methyl beta-bromopropionate, 3-bromopropionic acid methyl ester, propionic acid, 3-bromo-, methyl ester, 3-bromopropanoic acid methyl ester, unii-y803ch1p4w, beta-bromopropionic acid, methyl ester, methyl .beta.-bromopropionate, 3-bromo-propionic acid methyl ester
76934
methyl 3-bromopropanoate
COC(=O)CCBr

Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-bromopropionate was used in the synthesis of spiroanellated γ-lactones, pinacols and unprecedented 3-(1-hydroxycycloalkyl)-l-oxaspiro[n,m]alkan-2-ones. It can react with 2-Methyl-1H-benzoimidazole to get 1-(2-Methoxycarbonylethyl)-2-methylbenzimidazole.

Solubility
Immiscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.53
Boiling Point 58°C to 60°C (12 mmHg)
Flash Point 75°C (167°F)
Refractive Index 1.458
Quantity 25 g
Beilstein 1742949
Solubility Information Immiscible in water.
Formula Weight 167.01
Percent Purity 97%
Chemical Name or Material Methyl 3-bromopropionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'