Learn More
Methyl 3-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Catalog Number 15436907
Click to view available options
Quantity:
1 g
10 g
55279-30-6
C7H8N2O2
152.153
MFCD02082535
XLQIGLBALJNHKR-UHFFFAOYSA-N
1488824
methyl 3-aminopyridine-4-carboxylate
COC(=O)C1=C(C=NC=C1)N
55279-30-6
C7H8N2O2
152.153
MFCD02082535
XLQIGLBALJNHKR-UHFFFAOYSA-N
1488824
methyl 3-aminopyridine-4-carboxylate
COC(=O)C1=C(C=NC=C1)N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 84°C to 86°C
Quantity 1 g
Synonym methyl 3-aminoisonicotinate, 3-amino-isonicotinic acid methyl ester, 3-aminopyridine-4-carboxylic acid methyl ester, methyl3-aminoisonicotinate, 3-aminoisonicotinic acid methyl ester, 4-pyridinecarboxylic acid, 3-amino-, methyl ester, 3-amino-4-pyridinecarboxylic acid methyl ester, pubchem9760, acmc-1avm8, methyl 3-amino isonicotinate
Formula Weight 152.2
Percent Purity 97%
Chemical Name or Material Methyl 3-aminopyridine-4-carboxylate