Learn More
Methyl 3-amino-6-iodopyrazine-2-carboxylate, 96%, Thermo Scientific™
Product Code. 15499458
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15499458 5 g 5g
15408106 250 mg 250mg
15489458 1 g 1g
3 options
Product Code. 15499458
Brand: Thermo Scientific Alfa Aesar H63026.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1458-16-8
Molecular Formula C6H6IN3O2
Molecular Weight (g/mol) 279.037
MDL Number MFCD09909640
InChI Key FDLARAKNPMCCKJ-UHFFFAOYSA-N
Synonym 3-amino-6-iodopyrazine-2-carboxylic acid methyl ester, 3-amino-6-iodo-pyrazine-2-carboxylic acid methyl ester, methyl 3-amino-6-iodo-2-pyrazinecarboxylate, methyl 3-azanyl-6-iodanyl-pyrazine-2-carboxylate, 3-amino-6-iodo-2-pyrazinecarboxylic acid methyl ester, 2-pyrazinecarboxylic acid,3-amino-6-iodo-,methyl ester, 2-pyrazinecarboxylicacid, 3-amino-6-iodo-, methyl ester
PubChem CID 11087124
IUPAC Name methyl 3-amino-6-iodopyrazine-2-carboxylate
SMILES COC(=O)C1=NC(=CN=C1N)I
Specifications
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 279.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 3-amino-6-iodopyrazine-2-carboxylate