missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 11429127
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
51329-15-8
C8H6Br2O2
293.94
MFCD00082648
GSMAWUZTAIOCPL-UHFFFAOYSA-N
methyl3,5-dibromobenzoate, 3,5-dibromobenzoic acid methyl ester, benzoic acid, 3,5-dibromo-, methyl ester, 3,5-dibromomethylbenzoate, 1,3-dibromo-5-methoxycarbonyl benzene, pubchem3973, maybridge1_007706, acmc-209kt2, ksc274i7j, rarechem al bf 0770
621923
methyl 3,5-dibromobenzoate
COC(=O)C1=CC(Br)=CC(Br)=C1
This item is not returnable. View return policy
51329-15-8
C8H6Br2O2
293.94
MFCD00082648
GSMAWUZTAIOCPL-UHFFFAOYSA-N
methyl3,5-dibromobenzoate, 3,5-dibromobenzoic acid methyl ester, benzoic acid, 3,5-dibromo-, methyl ester, 3,5-dibromomethylbenzoate, 1,3-dibromo-5-methoxycarbonyl benzene, pubchem3973, maybridge1_007706, acmc-209kt2, ksc274i7j, rarechem al bf 0770
621923
methyl 3,5-dibromobenzoate
COC(=O)C1=CC(Br)=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 61°C to 64°C
Quantity 1 g
Beilstein 2255855
Formula Weight 293.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'