Learn More
Methyl 3,5-Dibromo-4-hydroxybenzoate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 3,5-Dibromo-4-hydroxybenzoic acid methyl ester |
| CAS | 41727-47-3 |
| Molecular Formula | C8 H6 Br2 O3 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C8H6Br2O3/c1-13-8(12)4-2-5(9)7(11)6(10)3-4/h2-3,11H,1H3 |
| SMILES | COC(=O)c1cc(Br)c(O)c(Br)c1 |
| IUPAC Name | methyl 3,5-dibromo-4-hydroxybenzoate |
| Molecular Weight (g/mol) | 309.94 |
| Formula Weight | 307.8684 |