Learn More
Methyl 3,4-dihydroxybenzoate, MedChemExpress
Product Code. 16475467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16475467 Supplier Medchem Express Supplier No. HYZ0548
 Restricted Item
Estimated Shipment: 01-12-2026
MedChemExpress Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl ester; Methyl protocatechuate) is a major metabolite of antioxidant polyphenols found in green tea. Antioxidant and anti-inflammatory effect.
Methyl 3,4-dihydroxybenzoate (Protocatechuic acid methyl ester
Specifications
Chemical Name or Material Methyl 3,4-dihydroxybenzoate
CAS 2150-43-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 50 mg
Synonym Protocatechuic acid methyl ester Methyl protocatechuate
Solubility Information DMSO : ≥ 100 mg/mL (594.71 mM)
SMILES O=C(OC)C1=CC=C(O)C(O)=C1
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 99.26%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death