missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4,5-trihydroxybenzoate, 99%

Product Code. 10616924 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 10616924

Brand: Thermo Scientific Acros 243462500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-24-1 | C8H8O5 | 184.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-24-1
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.15
MDL Number MFCD00002194
InChI Key FBSFWRHWHYMIOG-UHFFFAOYSA-N
Synonym methyl gallate, gallic acid methyl ester, methylgallate, benzoic acid, 3,4,5-trihydroxy-, methyl ester, gallic acid, methyl ester, methyl galloate, unii-623d3xg80c, ccris 5567, 3,4,5-trihydroxybenzoic acid methyl ester, chembl65675
PubChem CID 7428
IUPAC Name methyl 3,4,5-trihydroxybenzoate
SMILES COC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 199°C to 203°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 250 g
Merck Index 15, 4375
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol,methanol,ether
Formula Weight 184.15
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,4,5-trihydroxybenzoate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl 3,4,5-trihydroxybenzoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.