Learn More
Methyl 3-[2-Thienyl)propenoate, TRC
Product Code. 30472093
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472093 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472093 Supplier Toronto Research Chemicals Supplier No. TRCM330495250MG
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Chemical Name or Material Methyl 3-(2-Thienyl)propenoate
CAS 20883-96-9
Molecular Formula C8H8O2S
Quantity 250 mg
Synonym 2-Thiopheneacrylic Acid Methyl Ester, 3-(Thiophene-2-yl)acrylic Acid Methyl Ester
InChI Formula InChI=1S/C8H8O2S/c1-10-8(9)5-4-7-3-2-6-11-7/h2-6H,1H3/b5-4+
SMILES COC(=O)\C=C\c1cccs1
IUPAC Name methyl (E)-3-thiophen-2-ylprop-2-enoate
Molecular Weight (g/mol) 168.21
Formula Weight 168.0245
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.