Learn More
Methyl 2-(o-tolyl)acetate, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 40851-62-5 |
| Molecular Formula | C10H12O2 |
| MDL Number | MFCD00082780 |
| Quantity | 1 g |
| SMILES | O=C(OC)CC1=CC=CC=C1C |
| Molecular Weight (g/mol) | 164.20 |
| Percent Purity | 95% |
| Chemical Name or Material | Methyl 2-(o-tolyl)acetate |