Learn More
Methyl 2-Methoxy-5-(methylsulfonyl)benzoate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Methyl 2-Methoxy-5-(methylsulfonyl)benzoate
Specifications
Specifications
| Chemical Name or Material | Methyl 2-Methoxy-5-(methylsulfonyl)benzoate |
| CAS | 63484-12-8 |
| Analyte or Component Names | Methyl 2-Methoxy-5-(methylsulfonyl)benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Methoxy-5-(methylsulfonyl)benzoic acid methyl ester, Methyl 2-Methoxy-5-(methylsulfonyl)-benzoate, Tiapride Hydrochloride Imp. A (EP), Tiapride Hydrochloride Impurity A, Tiapride Impurity A |
| InChI Formula | InChI=1S/C10H12O5S/c1-14-9-5-4-7(16(3,12)13)6-8(9)10(11)15-2/h4-6H,1-3H3 |
| SMILES | COC(=O)c1cc(ccc1OC)S(=O)(=O)C |
| IUPAC Name | methyl 2-methoxy-5-methylsulfonylbenzoate |
| Show More |