Learn More
Methyl (2-furfurylthio)acetate, 97%, Thermo Scientific™
Product Code. 11485367
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485367 100 g 100g
11465367 5 g 5g
11475367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11485367 Supplier Thermo Scientific Alfa Aesar Supplier No. B22717.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108499-33-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD02283460
InChI Key XPVNCXPHDABNLE-UHFFFAOYSA-N
Synonym methyl 2-furfurylthio acetate, methyl 2-furan-2-ylmethyl thio acetate, acetic acid,2-2-furanylmethyl thio-, methyl ester, methyl 2-furan-2-ylmethyl sulfanyl acetate, acmc-20f6v3, aceticacid, 2-furanylmethyl thio-methylester, methyl-2-furfurylthio acetate, methyl 2-2-furylmethylthio acetate, furfurylthio acetic acid methyl ester, 2-furfurylthio acetic acid methyl ester
PubChem CID 7022533
IUPAC Name methyl 2-(furan-2-ylmethylsulfanyl)acetate
SMILES COC(=O)CSCC1=CC=CO1
Specifications
Boiling Point 95°C (3 mmHg)
Flash Point >100°C (212°F)
Quantity 100 g
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Methyl (2-furfurylthio)acetate