Learn More
Methyl 2-Butynoate, TRC
Product Code. 30469975
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469975 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30469975 Supplier Toronto Research Chemicals Supplier No. TRCM313745500MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-Butynoate
CAS 23326-27-4
Molecular Formula C5H6O2
Quantity 500 mg
Synonym Methyl 3-methylpropiolate, Methyl methylacetylenecarboxylate, Methyl methylpropiolate, Methyl tetrolateNSC 244950
InChI Formula InChI=1S/C5H6O2/c1-3-4-5(6)7-2/h1-2H3
SMILES COC(=O)C#CC
IUPAC Name methyl but-2-ynoate
Molecular Weight (g/mol) 98.1
Formula Weight 98.0368