Learn More
Methyl 2-Amino-5-methylthiophene-3-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 2-Amino-5-methylthiophene-3-carboxylate |
| CAS | 19369-53-0 |
| Molecular Formula | C7H9NO2S |
| Quantity | 2.5 g |
| Synonym | 2-Amino-5-methyl-3-thiophenecarboxylic Acid Methyl Ester, 2-Amino-3-(methoxycarbonyl)-5-methylthiophene, 2-Amino-5-methylthiophene-3-carboxylic Acid Methyl Ester |
| InChI Formula | InChI=1S/C7H9NO2S/c1-4-3-5(6(8)11-4)7(9)10-2/h3H,8H2,1-2H3 |
| SMILES | COC(=O)c1cc(C)sc1N |
| IUPAC Name | methyl 2-amino-5-methylthiophene-3-carboxylate |
| Molecular Weight (g/mol) | 171.22 |
| Formula Weight | 171.0354 |