Learn More
Methyl 2,4-difluorobenzoate, 98%, Thermo Scientific™
Product Code. 15454847
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15454847 5 g 5g
15464847 25 g 25g
2 options
Product Code. 15454847
Brand: Thermo Scientific Alfa Aesar H32161.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106614-28-2
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD03093799
InChI Key YQUHULOBTDYMAG-UHFFFAOYSA-N
Synonym 2,4-difluoro-benzoic acid methyl ester, 2,4-difluorobenzoic acid methyl ester, methyl-2,4-difluorobenzoate, methyl 2,4-difluoro benzoate, benzoic acid, 2,4-difluoro-, methyl ester, pubchem15678, acmc-1c4sx, methyl 2,4-diflourobenzoate, ksc494g3p, rarechem al bf 0213
PubChem CID 2782897
IUPAC Name methyl 2,4-difluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)F
Specifications
Quantity 5 g
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material Methyl 2,4-difluorobenzoate