Learn More
Methyl 2,3,3,3-Tetrafluoro-2-(1,1,2,3,3,3-hexafluoro-2-(perfluoropropoxy)propoxy)propanoate, TRC
Product Code. 30475295
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475295 50 mg 50mg
30472640 100 mg 100mg
30474904 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30475295 Supplier Toronto Research Chemicals Supplier No. TRCM21899850MG
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26131-32-8
Molecular Formula C10H3O4F17
Molecular Weight (g/mol) 510.1
Synonym Perfluoro-2,5-Dimethyl-3,6-Dioxanonanoic Acid Methyl Ester
IUPAC Name methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propanoate
SMILES COC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C10H3F17O4/c1-29-2(28)3(11,6(15,16)17)30-10(26,27)5(14,8(21,22)23)31-9(24,25)4(12,13)7(18,19)20/h1H3
Formula Weight 509.976
Chemical Name or Material Methyl 2,3,3,3-Tetrafluoro-2-(1,1,2,3,3,3-hexafluoro-2-(perfluoropropoxy)propoxy)propanoate