Learn More
Methyl 1-(2,4-Difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 1-(2,4-Difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate |
| CAS | 1202030-37-2 |
| Molecular Formula | C17 H12 F2 N2 O2 |
| Quantity | 100 mg |
| Synonym | 1H-Pyrazole-5-carboxylic acid, 1-(2,4-difluorophenyl)-3-phenyl-, methyl ester (ACI), Methyl 1-(2,4-difluorophenyl)-3-phenyl-1H-pyrazole-5-carboxylate (ACI) |
| InChI Formula | InChI=1S/C17H12F2N2O2/c1-23-17(22)16-10-14(11-5-3-2-4-6-11)20-21(16)15-8-7-12(18)9-13(15)19/h2-10H,1H3 |
| SMILES | COC(=O)c1cc(nn1c2ccc(F)cc2F)c3ccccc3 |
| IUPAC Name | methyl 2-(2,4-difluorophenyl)-5-phenyl-pyrazole-3-carboxylate |
| Molecular Weight (g/mol) | 314.29 |
| Formula Weight | 314.0867 |