missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Synonym | methoxyphenamine hydrochloride, methoxyphenamine hcl, orthoxine hydrochloride, ortodrinex hydrochloride, methoxyphenaminium chloride, methoxiphenadrin hydrochloride, o-methoxymethamphetamine hydrochloride, 1-2-methoxyphenyl propan-2-yl methyl amine hydrochloride, 1-2-methoxyphenyl-n-methylpropan-2-amine hydrochloride |
| SMILES | CC(CC1=CC=CC=C1OC)NC.Cl |
| IUPAC Name | 1-(2-methoxyphenyl)-N-methylpropan-2-amine;hydrochloride |
| PubChem CID | 9859211 |
| Formula Weight | 215.7 |
| Physical Form | Powder |
| Chemical Name or Material | Methoxyphenamine Hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?