missing translation for 'onlineSavingsMsg'
Learn More

Methoxyacetyl chloride, 97%, stabilized

Product Code. p-3778941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
Product Code. Quantity Packaging unitSize
10726052 10 g Glass bottle 10g
10703013 50 g Glass bottle 50g
10687304 250 g Glass bottle 250g
3 options
This item is not returnable. View return policy

Product Code. 10726052

Brand: Thermo Scientific Acros 125700100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38870-89-2 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38870-89-2
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000728
InChI Key JJKWHOSQTYYFAE-UHFFFAOYSA-N
Synonym methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
PubChem CID 96623
IUPAC Name 2-methoxyacetyl chloride
SMILES COCC(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1870g/mL
Boiling Point 112°C to 113°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OCH2COCl
Refractive Index 1.4190 to 1.422
Quantity 10 g
Fieser 12,310
Specific Gravity 1.187
Solubility Information Solubility in water: reacts
Stabilizer 0.3%mgO
Formula Weight 108.52
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methoxyacetyl chloride, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methoxyacetyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.