missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoprene-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Methoprene-d7 |
| Molecular Formula | C19 D7 H27 O3 |
| Quantity | 1 mg |
| Synonym | (2E,4E)-11-Methoxy-3,7,11-trimethyl-2,4-Dodecadienoic Acid 1-Methylethyl Ester-d7, (2E,4E)-3,7,11-Trimethyl-11-methoxydodeca-2,4-dienoic Acid Ethyl Ester-d7, Altosid-d7, Altosid Liquid Larvicide-d7, Altosid SR 10-d7, Altosid XR-d7, Apex-d7, Apex (pesticide)-d7, Apex 5E-d7, Apex 600E-d7, Bioprene BM Fire Ant Killer Bait-d7, Dianex-d7, ENT 70460-d7, Isopropyl (2E,4E)-11-methoxy-3,7,11-trimethyl-2,4-dodecadienoate-d7, Juvemon-d7, Juvenmon-d7, Kabat-d7, Manina-d7, Manta-d7, Manta (hormone)-d7, Methoprene S-d7, Metoprag 20CE-d7, SB-515-d7, Starbar Inhibitor-d7, ZPA 1019-d7, ZR 515-d7, dl-Isopropyl 11-Methoxy-3,7,11-trimethyl-trans-trans-2,4-dodecadienoate-d7 |
| InChI Formula | InChI=1S/C19H34O3/c1-15(2)22-18(20)14-17(4)11-8-10-16(3)12-9-13-19(5,6)21-7/h8,11,14-16H,9-10,12-13H2,1-7H3/b11-8+,17-14+/i1D3,2D3,15D |
| SMILES | [2H]C([2H])([2H])C([2H])(OC(=O)\C=C(/C)\C=C\CC(C)CCCC(C)(C)OC)C([2H])([2H])[2H] |
| IUPAC Name | [1,2,2,2-tetradeuterio-1-(trideuteriomethyl)ethyl] (2E,4E)-11-methoxy-3,7,11-trimethyl-dodeca-2,4-dienoate |
| Molecular Weight (g/mol) | 317.51 |
| Formula Weight | 317.2947 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?