Learn More
Method DM 471 Standard Mixture 888 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 71-43-2, 100-41-4, 1634-04-4, 108-38-3, 95-47-6, 106-42-3 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |