missing translation for 'onlineSavingsMsg'
Learn More

(±)-Methionine, TRC

Product Code. 30006284
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006284 100 g 100g
30006754 250 g 250g
30012895 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006284 Supplier Toronto Research Chemicals Supplier No. TRCM260395100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-51-8
InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)
IUPAC Name 2-amino-4-methylsulfanylbutanoic acid
Molecular Formula C5 H11 N O2 S
Molecular Weight (g/mol) 149.21
Chemical Name or Material DL-Methionine

Specifications

Synonym DL-Methionine, (2RS)-2-Amino-4-(methylsulfanyl)butanoic acid, Methionine, Methionine, DL- (8CI), (±)-Methionine, 2-Amino-4-(methylthio)butanoic acid, Acimetion, Amurex, Banthionine, Cynaron, DL-2-Amino-4-(methylthio)butyric acid, Dyprin, Lactet, Lobamine, Meonine, Meprom M 85, MetAMINO, Methilanin, MetiPEARL, Metione, NSC 9241, Neston, Pedameth, Racemethionine, Rhodimet NP 99, Smartamine, Smartamine M, Timet, Urimeth, α-Amino-γ-methylmercaptobutyric acid
SMILES CSCCC(N)C(=O)O
Quantity 100 g
Formula Weight 149.051
Percent Purity >95%
Shipping Condition Room Temperature
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.