Learn More
Methazolamide, Dr. Ehrenstorfer
Product Code. 30325030
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30325030 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325030 Supplier Dr. Ehrenstorfer Supplier No. DREC14999900
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methazolamide
CAS 554-57-4
Analyte or Component Names Methazolamide
Shipping Condition Room Temperature
Molecular Formula C5 H8 N4 O3 S2
Certifications/Compliance ISO/IEC 17025
Synonym N-[5-(aminosulfonyl)-3-methyl-1,3,4-thiadiazol-2(3H)-ylidene]acetamide;
InChI Formula InChI=1S/C5H8N4O3S2/c1-3(10)7-4-9(2)8-5(13-4)14(6,11)12/h1-2H3,(H2,6,11,12)/b7-4-
SMILES CN1N=C(S/C/1=N\C(=O)C)S(=O)(=O)N
IUPAC Name (NZ)-N-(3-methyl-5-sulfamoyl-1,3,4-thiadiazol-2-ylidene)acetamide
Molecular Weight (g/mol) 236.27
Quantity 50 mg
Formula Weight 236.0038 g/mol