Learn More
Methanol, Supelco™
Product Code. 15804730
Change view
Click to view available options
Quantity:
2.5 L
10 L
30 L
185 L
Unit Size:
10 L
185 L
2.5 L
30 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15804730 2.5 L 2.5 L
15814730 10 L 10 L
15824730 30 L 30 L
15807420 185 L 185 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15804730 Supplier Supelco Supplier No. 1060072500
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
In Stock
Gradient grade for liquid chromatography LiChrosolv™ Reag. Ph Eur, Reag. ChP
LiChrosolv™ solvents were specially formulated to meet the demanding needs of analytical HPLC applications. With exceptional UV transmittance, minimal particle count, and controlled acidity and alkalinity, our solvents ensure reproducible separations every time. The low evaporation residue level further enhances their performance, making them the ideal choice for high-quality results. The product line includes both gradient grade and isocratic grade solvents, specifically designed to reduce the gradient effect during separations. This tailored approach allows for optimal consistency and accuracy in your analytical processes, helping you achieve the best possible outcomes in your chromatography work. Now Available with Data Matrix Code This high-quality HPLC-grade solvent now features a unique Data Matrix Code on its label, providing digital access to essential product information such as batch number, product number, and expiry date. Unlike standard barcodes, our DMCs feature a unique serial number for each bottle, enabling you to monitor and trace every bottle individually. With standard barcodes, all bottles from the same batch look identical. Our solution gives you true bottle-level traceability for greater control and compliance in the lab. Know more about our data matrix code-labeled HPLC-grade solvents.
TRUSTED_SUSTAINABILITY
Specifications
Density 0.791 g/mL at 25°C
Quantity 2.5 L
Molecular Weight (g/mol) 32.04
compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Methanol gradient grade for liquid chromatography LiChrosolv® Reag. Ph Eur,Reag. ChP
Avvertenza
  • Danger
Categoria di pericolo
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Indicazioni di pericolo
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.
Consigli di prudenza
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P311-Call a POISON CENTER/doctor/ .
  • P330-Rinse mouth.
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.