missing translation for 'onlineSavingsMsg'
Learn More

Methanol - D4, deuteration degree min. 99.8 % for NMR spectroscopy MagniSolv™, MilliporeSigma™

Product Code. 15894750
Change view
Click to view available options
Quantity:
10 mL
25 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
Unit Size:
10mL
25mL
7.5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15894750 10 mL Septum Ampule 10mL
15884750 10 x 0.75 mL Glass Ampule 7.5mL
15804760 25 mL Glass Bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15894750 Supplier Sigma-Aldrich Supplier No. 1060280010
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Odor Characteristic
Packaging Septum Ampule
Vapor Pressure 128 hPa at 20°C (68°F)
Viscosity 0.597mPa.s (20°C/68°F)
Quantity 10 mL
Assay Percent Range 99.8% min.
UN Number 1230
Formula Weight 36.07
Chemical Name or Material Methanol - D4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.