Learn More
Methanol - D4, deuteration degree min. 99.8 % for NMR spectroscopy MagniSolv™, MilliporeSigma™
Product Code. 15804760
Change view
Click to view available options
Quantity:
10 mL
25 mL
10 x 0.75 mL
Packaging:
Glass Ampule
Glass Bottle
Septum Ampule
Unit Size:
10mL
25mL
7.5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15804760 25 mL Glass Bottle 25mL
15884750 10 x 0.75 mL Glass Ampule 7.5mL
15894750 10 mL Septum Ampule 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15804760 Supplier Sigma Aldrich Supplier No. 1060280025
 Restricted Item
Estimated Shipment: 18-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
For NMR Spectroscopy
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO
Specifications
Physical Form Liquid
Color Colorless
Density 0.79g/cm3 at 20°C (68°F)
Flash Point 11°C
Odor Characteristic
Packaging Glass Bottle
Vapor Pressure 128 hPa at 20°C (68°F)
Viscosity 0.597mPa.s (20°C/68°F)
Quantity 25 mL
Melting Point -98°C (1013hPa)
Boiling Point 64.5°C
Assay Percent Range 99.8% min.
UN Number 1230
Formula Weight 36.07
Chemical Name or Material Methanol - d4