Learn More
SPEX CertiPrep™ Methacrylate Grinding Balls
Product Code. 12578706 Shop All SPEX Certiprep Products
Change view
Click to view available options
Diameter (Metric):
9.5 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) unitSize
12578706 9.5 mm Pack of 100
1 options
Product Code. 12578706
Brand: SPEX CertiPrep™ 3112
This item requires a price update prior to being ordered.
Request a Price for this item here.
Disposable grinding ball ideal for pulverizing soft, brittle materials and for mixing powders.
  • Typical composition: Major Element – C
  • Hardness: Mohs 1 ½
  • Compatible with all SPEX SamplePrep plastic grinding vials.
Specifications
Diameter (English) 3/8 in.
Diameter (Metric) 9.5 mm
For Use With (Equipment) SPEX SamplePrep plastic vials
Material Methacrylate
Packaging Quantity 100/bag