Learn More
Methacrifos, Dr. Ehrenstorfer
Product Code. 30322148
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 30322148
Brand: Dr. Ehrenstorfer DREC14970000
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methacrifos
CAS 62610-77-9
Analyte or Component Names Methacrifos
Shipping Condition Room Temperature
Molecular Formula C7 H13 O5 P S
Certifications/Compliance ISO 17034
Synonym 2-Propenoic acid, 3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-, methyl ester, (2E)-, 2-Propenoic acid, 3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-, methyl ester, (E)-, CGA 20168, trans-Methacrifos, Methacrifos
InChI Formula InChI=1S/C7H13O5PS/c1-6(7(8)9-2)5-12-13(14,10-3)11-4/h5H,1-4H3/b6-5+
Solution Type Neat
SMILES COC(=O)\C(=C\OP(=S)(OC)OC)\C
IUPAC Name methyl (E)-3-dimethoxyphosphinothioyloxy-2-methylprop-2-enoate
Industry Type Environmental Testing
Molecular Weight (g/mol) 240.21
Quantity 100 mg
Formula Weight 240.0221 g/mol