Learn More
Metamitron, TRC
Product Code. 30003263
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003263 5 g 5g
30011060 1 g 1g
30003225 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003263 Supplier Toronto Research Chemicals Supplier No. TRCM2588435G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41394-05-2
Molecular Formula C10 H10 N4 O
Molecular Weight (g/mol) 202.21
Synonym 4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5(4H)-one, 3-Methyl-4-amino-6-phenyl-1,2,4-triazin-5-one, 4-Amino-3-methyl-6-phenyl-1,2,4-triazin-5-one, BAY 6676, BAY-DRW 1139, Burakomitron, DRW 1139, Gladiator, Goltix, Goltix Compact, Goltix Top, Goltix VM 90, Goltix WG, Goltix WG 70, Metafol 700SC, Metamitron, Metron 700SL, Metron Stefes
IUPAC Name 4-amino-3-methyl-6-phenyl-1,2,4-triazin-5-one
SMILES CC1=NN=C(C(=O)N1N)c2ccccc2
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H10N4O/c1-7-12-13-9(10(15)14(7)11)8-5-3-2-4-6-8/h2-6H,11H2,1H3
Formula Weight 202.0855
Chemical Name or Material Metamitron