Learn More
Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes
Product Code. 12996986 Shop All Nickel-Electro Products
Change view
Click to view available options
Length (Metric):
300 mm
340 mm
400 mm
430 mm
490 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
12996986 400 mm Each
11844573 300 mm Each
12966986 340 mm Each
12976986 430 mm Each
12986986 490 mm Each
5 options
Product Code. 12996986
Brand: Nickel Electro™ 6706/SS
Only null left
Add to basket
Add to basket
Keep pipets sterile with a sterilizing box. Nickel Electro™ Metalware Range™ Stainless Steel Square Pipet Sterilizing Boxes can be steam sterilized at 120°C and withstand dry heat temperatures of up to 180°C.
  • Stainless steel square profile
  • Includes lid
  • Square shape prevents it from rolling off bench top
  • Silicon in lid and base
Specifications
Autoclavable Autoclavable
Dimensions (L x W) 400 x 70 mm
Material Stainless Steel
Sterility Sterile
Quantity Each
Length (Metric) 400 mm
Type Sterilizing Box
For Use With (Equipment) Autoclave