missing translation for 'onlineSavingsMsg'
Learn More
Learn More
metallothionein Antibody (1F5) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18261JF585
This item is not returnable.
View return policy
Description
metallothionein Monoclonal antibody specifically detects metallothionein in Bacteria samples. It is validated for ELISA,Western Blot
Specifications
| metallothionein | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
| ELISA, Western Blot | |
| 1F5 | |
| 50mM Sodium Borate | |
| Recombinant full length Pseudomonas aeruginosa Metallothionein protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction