Learn More
Paraben target analyte mix solution, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 67-64-1 |
| Density | 0.79 g/cm3 |
| Flash Point | -17°C |
| Physical Form | Liquid |
| Boiling Point | 55°C to 57°C |
| Industry Type | Parabenes |
| Quantity | 1 mL |
| Grade | Analytical Standard |