Learn More
Brady™ MetaLabel™ B-486 Metallized Polyester Labels with an Aggressive Adhesive
Click to view available options
Size (English):
1 x 0.5 in. (W x H)
1.5 x 0.5 in. (W x H)
1.5 x 0.75 in. (W x H)
2 x 1 in. (W x H)
3 x 1.9 in. (W x H)
Unit Size:
Each
Specifications
Specifications
| Certifications/Compliance | UL Recognized, CSA Approved |
| Description | Chemical, Corrosion, Humidity, UV and Weather-Resistant |
| Adhesive Type | Rubber, Permanent |
| Color | Light Gray |
| Finish | Matte |
| Material | Metallized Polyester (B-486) |
| Product Type | Asset Tracking Label |
| For Use With (Application) | Asset tracking, Rating plates, General purpose ID |
| For Use With (Equipment) | BMP™71 Printer |
| Quantity | 100/Pk. |
| Show More |