Learn More
Brady™ Metal Wall Lock Box
Product Code. p-7073793
Change view
Click to view available options
Includes:
(7) key hooks
Dimensions (L x W x H):
2.5 x 8 x 7 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Dimensions (L x W x H) unitSize
15611127 (7) key hooks 2.5 x 8 x 7 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15611127 Supplier Brady™ Supplier No. 105714
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Metal Wall Lock Box is made of durable, powder-coated steel with key slot on top. Includes clear Lexan window that slides out when locks are removed.
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Product Type Wall Mount Lock Box
Color Yellow on Red
Material Powder-coated Steel
Dimensions (L x W x H) 2.5 x 8 x 7 in.
Displayed Text GROUP LOCK BOX
Height (English) 7 in.
Includes (7) key hooks
Length (English) 2.5 in.
Quantity 1 Ea.
Width (English) 8 in.