Learn More
VACUUBRAND™ Metal Vacuum Tubing With Small Flanges KF
Description
- Manufactured from stainless steel
- Available in a range of sizes and lengths
Specifications
Specifications
| Description | Single bending radius of 43 |
| Diameter (Metric) Inner | 25 mm |
| Material | Metal |
| Overall Length (Metric) | 75 cm |
| Packaging Quantity | 1 Each |
| Tubing Size | 25 |
| Product Type | Vacuum Tubing |