Learn More
Sampling Systems™ Metal Detectable Long Handled Scoops
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 4
Description
- Our Long Handled Metal Detectable Scoops are perfect for both sampling and dispensing when working with powders and granules.
- The long handle and the bright blue color make the scoops easy to spot if they get left behind.
- To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom.
- The robust nature of these scoops means that they can give many years of service or, for more demanding applications, they can be used as a single use disposable product straight from the bag.
- The scoops are made from polypropylene that has been mixed with a special food grade filler.
- It is the filler that makes the scoops metal detectable and X-Ray visible.
- The filler is uniformly distributed throughout the scoop so even small fragments of scoop can still be detected.
- Simply unwrap and use.
- These scoops are ideal for use with powders, granules and pastes.
- Our Metal Detectable Long Handle Scoops are designed to be as easy to clean as possible.
- The main body of the scoop has rounded corners.
- Made from food grade PP.
- Conforms to the latest FDA and EU requirements.
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Color | Blue |
| Material | Polypropylene |