Learn More
Pyrimethanil in Methanol 100μg/mL, Fisher Chemical™
Product Code. 12996533 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 12996533
Brand: Fisher Chemical P850750
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule
CAS 53112-28-0
Physical Form Liquid
Molecular Formula C12H13N3
Concentration 100 μg/mL
InChI Key ZLIBICFPKPWGIZ-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CC1=CC(=NC(=N1)NC2=CC=CC=C2)C
IUPAC Name 4,6-dimethyl-N-phenylpyrimidin-2-amine
Solvent or Matrix Methanol
Molecular Weight (g/mol) 199.257
PubChem CID 91650
Quantity 1 mL
ChEBI CHEBI:8674
Chemical Name or Material Pyrimethanil