Learn More
MCPA in Methanol 10μg/mL, Fisher Chemical™
Product Code. 12979873 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. 12979873
Brand: Fisher Chemical P830580
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 94-74-6
Molecular Formula C9H9ClO3
Concentration 10 μg/mL
UN Number 1230
InChI Key WHKUVVPPKQRRBV-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Cl)OCC(=O)O
IUPAC Name 2-(4-chloro-2-methylphenoxy)acetic acid
Solvent or Matrix Methanol
Molecular Weight (g/mol) 200.618
PubChem CID 7204
Quantity 10 mL
ChEBI CHEBI:50099
Chemical Name or Material MCPA
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution MCPA 10ug/ml in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .