Learn More
Hexachlorobenzene in n-Nonane 100μg/mL, Fisher Chemical™
Product Code. 12996863 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12996863 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12996863 Supplier Fisher Chemical Supplier No. P828310
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 93952-14-8
Molecular Formula C6Cl6
Concentration 100 μg/mL
InChI Key CKAPSXZOOQJIBF-IDEBNGHGSA-N
SMILES C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)Cl
IUPAC Name 1,2,3,4,5,6-hexachlorobenzene
Solvent or Matrix n-Nonane
Molecular Weight (g/mol) 290.72
PubChem CID 12214812
Quantity 1 mL
Chemical Name or Material Hexachlorobenzene 13C6